Merck
  • [Screening of endophytic fungi from Huperzia serrata for acetylcholinesterase inhibitory activity and its taxonomic identification].
[Screening of endophytic fungi from Huperzia serrata for acetylcholinesterase inhibitory activity and its taxonomic identification].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2013-05-01)
Li-Li Wang, Hui-Fang Lv, Li Zhang, Hai-Xia Hua, Jie-Hua Wang, Zhi-Bi Hu, Wan-Kui Li
ABSTRACT
To screen out fungus strains with acetylcholinesterase inhibitory activity from Huperzia serrata. Endophytic fungi fermentation products from 59 H. serrata strains were stained with acetylcholinesterase hydrolyzed alpha-naphthaleneacetic ethyl ester and fast blue B salt, and screened for acetylcholinesterase inhibitory activity with thin-layer chromatography-bioautography. Target strains were classified and identified through the sequence analysis on 18s rDNA and 5.8s rDNA combined with morphological characteristics. Fungus strain LQ2F01 from H. serrata showed positive color reaction in the screening for acetylcholinesterase inhibitory activity. The sequence analysis on 18s rDNA and 5.8s rDNA combined with morphological characteristics showed the strain LQ2F01 belonged to Acremonium. Endophytic Fungi LQ2F01 from H. serrata shows identical acetylcholinesterase inhibitory activity with the host plant, which is of great significance to the development of natural medicines and the studies on the relationship between the endophytic gungi and the host plant.
Product Description
Supelco
1-Naphthaleneacetic acid, PESTANAL®, analytical standard
Sigma-Aldrich
1-Naphthaleneacetic acid, technical grade
Sigma-Aldrich
Acetylcholinesterase human, recombinant, expressed in HEK 293 cells, lyophilized powder, ≥1,000 units/mg protein (Lowry)
Sigma-Aldrich
1-Naphthaleneacetic acid, suitable for plant cell culture, BioReagent, ≥95%, crystalline
Sigma-Aldrich
Acetylcholinesterase from Electrophorus electricus (electric eel), Type V-S, lyophilized powder, ≥1,000 units/mg protein
Sigma-Aldrich
Fast Blue BB Salt hemi(zinc chloride) salt, Dye content ≥80 %
Sigma-Aldrich
1-Naphthylacetic acid, BioReagent, 1 mg/mL, suitable for plant cell culture
Sigma-Aldrich
Acetylcholinesterase from Electrophorus electricus (electric eel), Type VI-S, lyophilized powder, 200-1,000 units/mg protein
Sigma-Aldrich
Acetylcholinesterase from human erythrocytes, buffered aqueous solution, ≥500 units/mg protein (BCA)
Sigma-Aldrich
Fast Blue BB Salt hemi(zinc chloride) salt, for microscopy (Hist.)