Merck
The GUS reporter gene system.
Nature (1989-12-14)
R A Jefferson
ABSTRACT
The GUS reporter gene system is already a powerful tool for the assessment of gene activity in transgenic plants. Further developments may lead to routine in vivo analysis and fusion genetics.
Sigma-Aldrich
β-Glucuronidase Reporter Gene Staining Kit