Merck
  • Prognostic and predictive significance of MYC and KRAS alterations in breast cancer from women treated with neoadjuvant chemotherapy.
Prognostic and predictive significance of MYC and KRAS alterations in breast cancer from women treated with neoadjuvant chemotherapy.
PloS one (2013-04-05)
Cynthia Brito Lins Pereira, Mariana Ferreira Leal, Carolina Rosal Teixeira de Souza, Raquel Carvalho Montenegro, Juan Antonio Rey, Antônio Alberto Carvalho, Paulo Pimentel Assumpção, André Salim Khayat, Giovanny Rebouças Pinto, Sâmia Demachki, Marília de Arruda Cardoso Smith, Rommel Rodríguez Burbano
ABSTRACT
Breast cancer is a complex disease, with heterogeneous clinical evolution. Several analyses have been performed to identify the risk factors for breast cancer progression and the patients who respond best to a specific treatment. We aimed to evaluate whether the hormone receptor expression, HER2 and MYC genes and their protein status, and KRAS codon 12 mutations may be prognostic or predictive biomarkers of breast cancer. Protein, gene and mutation status were concomitantly evaluated in 116 breast tumors from women who underwent neoadjuvant chemotherapy with doxorubicin plus cyclophosphamide. We observed that MYC expression was associated with luminal B and HER2 overexpression phenotypes compared to luminal A (p<0.05). The presence of MYC duplication or polysomy 8, as well as KRAS mutation, were also associated with the HER2 overexpression subtype (p<0.05). MYC expression and MYC gain were more frequently observed in early-onset compared to late-onset tumors (p<0.05). KRAS mutation was a risk factor of grade 3 tumors (p<0.05). A multivariate logistic regression demonstrated that MYC amplification defined as MYC/nucleus ratio of ≥2.5 was a protective factor for chemotherapy resistance. On the other hand, age and grade 2 tumors were a risk factor. Additionally, luminal B, HER2 overexpression, and triple-negative tumors presented increased odds of being resistant to chemotherapy relative to luminal A tumors. Thus, breast tumors with KRAS codon 12 mutations seem to present a worse prognosis. Additionally, MYC amplification may help in the identification of tumors that are sensitive to doxorubicin plus cyclophosphamide treatment. If confirmed in a large set of samples, these markers may be useful for clinical stratification and prognosis.
Sigma-Aldrich
Anti-Estrogen Receptor-α antibody produced in rabbit, affinity isolated antibody