Merck
  • Cuprous oxide nanoparticles dispersed on reduced graphene oxide as an efficient electrocatalyst for oxygen reduction reaction.
Cuprous oxide nanoparticles dispersed on reduced graphene oxide as an efficient electrocatalyst for oxygen reduction reaction.
Chemical communications (Cambridge, England) (2012-01-10)
Xiao-Yan Yan, Xi-Li Tong, Yue-Fei Zhang, Xiao-Dong Han, Ying-Yong Wang, Guo-Qiang Jin, Yong Qin, Xiang-Yun Guo
ABSTRACT
Cuprous oxide (Cu(2)O) nanoparticles dispersed on reduced graphene oxide (RGO) were prepared by reducing copper acetate supported on graphite oxide using diethylene glycol as both solvent and reducing agent. The Cu(2)O/RGO composite exhibits excellent catalytic activity and remarkable tolerance to methanol and CO in the oxygen reduction reaction.
Sigma-Aldrich
Diethylene glycol, BioUltra, ≥99.0% (GC)