Merck
  • The Streptomyces-produced antibiotic fosfomycin is a promiscuous substrate for archaeal isopentenyl phosphate kinase.
The Streptomyces-produced antibiotic fosfomycin is a promiscuous substrate for archaeal isopentenyl phosphate kinase.
Biochemistry (2011-12-14)
Mark F Mabanglo, Adrian W R Serohijos, C Dale Poulter
ABSTRACT
Isopentenyl phosphate kinase (IPK) catalyzes the phosphorylation of isopentenyl phosphate to form the isoprenoid precursor isopentenyl diphosphate in the archaeal mevalonate pathway. This enzyme is highly homologous to fosfomycin kinase (FomA), an antibiotic resistance enzyme found in a few strains of Streptomyces and Pseudomonas whose mode of action is inactivation by phosphorylation. Superposition of Thermoplasma acidophilum (THA) IPK and FomA structures aligns their respective substrates and catalytic residues, including H50 and K14 in THA IPK and H58 and K18 in Streptomyces wedmorensis FomA. These residues are conserved only in the IPK and FomA members of the phosphate subdivision of the amino acid kinase family. We measured the fosfomycin kinase activity of THA IPK [K(m) = 15.1 ± 1.0 mM, and k(cat) = (4.0 ± 0.1) × 10⁻² s⁻¹], resulting in a catalytic efficiency (k(cat)/K(m) = 2.6 M⁻¹ s⁻¹) that is 5 orders of magnitude lower than that of the native reaction. Fosfomycin is a competitive inhibitor of IPK (K(i) = 3.6 ± 0.2 mM). Molecular dynamics simulation of the IPK·fosfomycin·MgATP complex identified two binding poses for fosfomycin in the IP binding site, one of which results in a complex analogous to the native IPK·IP·ATP complex that engages H50 and the lysine triangle formed by K5, K14, and K205. The other binding pose leads to a dead-end complex that engages K204 near the IP binding site to bind fosfomycin. Our findings suggest a mechanism for acquisition of FomA-based antibiotic resistance in fosfomycin-producing organisms.
Sigma-Aldrich
Isopentenyl phosphate dilithium salt, ≥95.0% (TLC)