Merck
  • Acceptor engineering of metallacycles with high phototoxicity indices for safe and effective photodynamic therapy.
Acceptor engineering of metallacycles with high phototoxicity indices for safe and effective photodynamic therapy.
Chemical science (2023-03-21)
Chonglu Li, Le Tu, Jingfang Yang, Chang Liu, Yuling Xu, Junrong Li, Wei Tuo, Bogdan Olenyuk, Yan Sun, Peter J Stang, Yao Sun
ABSTRACT
Although metallacycle-based photosensitizers have attracted increasing attention in biomedicine, their clinical application has been hindered by their inherent dark toxicity and unsatisfactory phototherapeutic efficiency. Herein, we employ a π-expansion strategy for ruthenium acceptors to develop a series of Ru(ii) metallacycles (Ru1-Ru4), while simultaneously reducing dark toxicity and enhancing phototoxicity, thus obtaining a high phototoxicity index (PI). These metallacycles enable deep-tissue (∼7 mm) fluorescence imaging and reactive oxygen species (ROS) production and exhibit remarkable anti-tumor activity even under hypoxic conditions. Notably, Ru4 has the lowest dark toxicity, highest ROS generation ability and an optimal PI (∼146). Theoretical calculations verify that Ru4 exhibits the largest steric bulk and the lowest singlet-triplet energy gap (ΔE ST, 0.62 eV). In vivo studies confirm that Ru4 allows for effective and safe phototherapy against A549 tumors. This work thus is expected to open a new avenue for the design of high-performance metal-based photosensitizers for potential clinical applications.
Product Description
Dichloro(p-cymene)ruthenium(II) dimer ChemBeads