Merck
  • Specific detection of Salmonella enterica and Escherichia coli strains by using ELISA with bacteriophages as recognition agents.
Specific detection of Salmonella enterica and Escherichia coli strains by using ELISA with bacteriophages as recognition agents.
European journal of clinical microbiology & infectious diseases : official publication of the European Society of Clinical Microbiology (2011-02-15)
E Galikowska, D Kunikowska, E Tokarska-Pietrzak, H Dziadziuszko, J M Loś, P Golec, G Węgrzyn, M Loś
ABSTRACT
The use of bacteriophages, instead of antibodies, in the ELISA-based detection of bacterial strains was tested. This procedure appeared to be efficient, and specific strains of Salmonella enterica and Escherichia coli could be detected. The sensitivity of the assay was about 10(5) bacterial cells/well (10(6)/ml), which is comparable with or outperforms other ELISA tests detecting intact bacterial cells without an enrichment step. The specificity of the assay depends on the kind of bacteriophage used. We conclude that the use of bacteriophages in the detection and identification of bacteria by an ELISA-based method can be an alternative to the use of specific antibodies. The advantages of the use of bacteriophages are their environmental abundance (and, thus, a possibility to isolate various phages with different specificities) and the availability of methods for obtaining large amounts of phage lysates, which are simple, rapid, cheap, and easy.
Sigma-Aldrich
ExtrAvidin® –Alkaline Phosphatase, buffered aqueous solution
Sigma-Aldrich
Alkaline Phosphatase Yellow (pNPP) Liquid Substrate System for ELISA, ready to use solution