Merck
  • MAP7 drives EMT and cisplatin resistance in ovarian cancer via wnt/β-catenin signaling.
MAP7 drives EMT and cisplatin resistance in ovarian cancer via wnt/β-catenin signaling.
Heliyon (2024-05-10)
Qingqing Chen, Shaojing Li, Furong Fu, Qunhuan Huang, Rong Zhang
ABSTRACT
Our approach encompasses analyzing MAP7's expression levels across various datasets and clinical specimens, evaluating its association with patient outcomes, and probing its influence on ovarian cancer cell dynamics such as proliferation, migration, invasion, and apoptosis. We have identified significant upregulation of MAP7 in ovarian cancer tissues, which correlates with advanced disease stages, higher pathological grades, and unfavorable prognoses. Functionally, the inhibition of MAP7 suppresses cancer cell proliferation, migration, and invasion while promoting apoptosis. Notably, the silencing of MAP7 attenuates the epithelial-mesenchymal transition (EMT) and disrupts Wnt/β-catenin pathway signaling-two critical processes implicated in metastasis and chemoresistance. In cisplatin-resistant A2780-DDP cells, the downregulation of MAP7 effectively reverses their resistance to cisplatin. Furthermore, the nuclear localization of MAP7 in these cells underscores its pivotal role in driving cisplatin resistance by modulating the transcriptional regulation and interaction dynamics of β-catenin. Our findings position MAP7 as a pivotal element in ovarian cancer advancement and cisplatin resistance, primarily through its modulation of EMT and the Wnt/β-catenin pathway. Its association with poor clinical outcomes underscores its potential as both a prognostic marker and a therapeutic target. Strategies aimed at MAP7 could represent a new frontier in combating chemotherapy resistance in ovarian cancer, emphasizing its significance in crafting complementary treatments for this disease.
Sigma-Aldrich
Anti-MAP7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1