Merck
  • Single-cell and bulk RNA sequencing reveal cancer-associated fibroblast heterogeneity and a prognostic signature in prostate cancer.
Single-cell and bulk RNA sequencing reveal cancer-associated fibroblast heterogeneity and a prognostic signature in prostate cancer.
Medicine (2023-08-11)
Wen Liu, Miaomiao Wang, Miao Wang, Ming Liu
ABSTRACT
Cancer-associated fibroblasts (CAFs), the central players in the tumor microenvironment (TME), can promote tumor progression and metastasis via various functions. However, the properties of CAFs in prostate cancer (PCa) have not been fully assessed. Therefore, we aimed to examine the CAF characteristics in PCa and construct a CAF-derived signature to predict PCa prognosis. CAFs were identified using single-cell RNA sequencing (scRNA-seq) data from 3 studies. We performed the FindAllMarkers function to extract CAF marker genes and constructed a signature to predict the biochemical relapse-free survival (bRFS) of PCa in the Cancer Genome Atlas (TCGA) cohort. Subsequently, different algorithms were applied to reveal the differences of the TME, immune infiltration, treatment responses in the high- and low-risk groups. Additionally, the CAF heterogeneity was assessed in PCa, which were confirmed by the functional enrichment analysis, gene set enrichment analysis (GSEA), and AUCell method. The scRNA-seq analysis identified a CAF cluster with 783 cells and determined 183 CAF marker genes. Cell-cell communication revealed extensive interactions between fibroblasts and immune cells. A CAF-related prognostic model, containing 7 genes (ASPN, AEBP1, ALDH1A1, BGN, COL1A1, PAGE4 and RASD1), was developed to predict bRFS and validated by 4 independent bulk RNA-seq cohorts. Moreover, the high-risk group of the signature score connected with an immunosuppressive TME, such as a higher level of M2 macrophages and lower levels of plasma cells and CD8+ T cells, and a reduced reaction rate for immunotherapy compared with low-risk group. After re-clustering CAFs via unsupervised clustering, we revealed 3 biologically distinct CAF subsets, namely myofibroblast-like CAFs (myCAFs), immune and inflammatory CAFs (iCAFs) and antigen-presenting CAFs (apCAFs). In conclusion, the CAF-derived signature, the first of its kind, can effectively predict PCa prognosis and serve as an indicator for immunotherapy. Furthermore, our study identified 3 CAF subpopulations with distinct functions in PCa.
Sigma-Aldrich
Anti-PAGE4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-AEBP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution