Merck
  • Targeted mutagenesis of the herpesvirus fusogen central helix captures transition states.
Targeted mutagenesis of the herpesvirus fusogen central helix captures transition states.
Nature communications (2023-12-03)
Momei Zhou, Benjamin Vollmer, Emily Machala, Muyuan Chen, Kay Grünewald, Ann M Arvin, Wah Chiu, Stefan L Oliver
ABSTRACT
Herpesviruses remain a burden for animal and human health, including the medically important varicella-zoster virus (VZV). Membrane fusion mediated by conserved core glycoproteins, the fusogen gB and the heterodimer gH-gL, enables herpesvirus cell entry. The ectodomain of gB orthologs has five domains and is proposed to transition from a prefusion to postfusion conformation but the functional relevance of the domains for this transition remains poorly defined. Here we describe structure-function studies of the VZV gB DIII central helix targeting residues 526EHV528. Critically, a H527P mutation captures gB in a prefusion conformation as determined by cryo-EM, a loss of membrane fusion in a virus free assay, and failure of recombinant VZV to spread in cell monolayers. Importantly, two predominant cryo-EM structures of gB[H527P] are identified by 3D classification and focused refinement, suggesting they represented gB conformations in transition. These studies reveal gB DIII as a critical element for herpesvirus gB fusion function.
Sigma-Aldrich
Anti-Varicella-Zoster Virus Antibody, Immediate Early Gene 62, Chemicon®, from mouse