Merck
  • Longitudinal analysis of anti-SARS-CoV-2 neutralizing antibody (NAb) titers in vaccinees using a novel giant magnetoresistive (GMR) assay.
Longitudinal analysis of anti-SARS-CoV-2 neutralizing antibody (NAb) titers in vaccinees using a novel giant magnetoresistive (GMR) assay.
Sensors and actuators. B, Chemical (2023-04-15)
Elaine Ng, Christopher Choi, Shan X Wang
ABSTRACT
The COVID-19 pandemic has highlighted the need to monitor important correlates of immunity on a population-wide level. To this end, we have developed a competitive assay to assess neutralizing antibody (NAb) titer on the giant magnetoresistive (GMR) biosensor platform. We compared the clinical performance of our biosensor with established techniques such as Ortho's VITROS Anti-SARS-CoV-2 IgG Quantitative Antibody test. Results obtained between the VITROS test and the GMR assay showed correlation (r = -0.93). We then validated the assay with patient plasma samples that had been tested using focus reduction neutralization testing (FRNT). The results obtained from our GMR assay exhibit a previously identified trend of increased NAb titers 2 weeks post-vaccination. We further evaluated NAb titers 6 months post-vaccination and observed waning neutralizing antibody titers over that time in vaccinated patients. In addition, we calibrated our assay to an arbitrary unit (IU/mL) using World Health Organization (WHO) reference plasma provided by the National Institute of Biological Standards and Control (NIBSC). Our biosensor provides highly specific and sensitive results in serum and plasma with analytical, clinical, and point-of-care (POC) applications due to quick turnaround times on samples and the cost-effectiveness of the platform.
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin, biotin labeled bovine, lyophilized powder