Merck
  • Thiol-Rich Multifunctional Macromolecular Crosslinker for Gelatin-Norbornene-Based Bioprinting.
Thiol-Rich Multifunctional Macromolecular Crosslinker for Gelatin-Norbornene-Based Bioprinting.
Biomacromolecules (2021-06-01)
Cailing Zhao, Zejia Wu, Hanyu Chu, Tao Wang, Shuai Qiu, Jing Zhou, Qingtang Zhu, Xiaolin Liu, Daping Quan, Ying Bai
ABSTRACT
Extrusion-based bioprinting is an emerging and most frequently used technique for the fabrication of cell-laden constructs. A suitable hydrogel-based bioink for cell encapsulation and protection is critical for printability, structural stability, and post-printing cell viability. The thiol-ene chemistry-based gelatin-norbornene (GelNB) hydrogels have drawn much attention as a promising substitution of gelatin methacryloyl (GelMA), owing to the fast and controllable step-growth polymerization mechanism, as well as a significant reduction in reactive oxygen species (ROS) accumulation. Herein, thiolated heparin (HepSH) was synthesized and used as a macromolecular crosslinker for GelNB-based bioprinting, so that GelNB gelation became less sensitive to the thiol/ene ratio. The mechanical stability and moduli of GelNB/HepSH hydrogels were easily manipulated by the concentration and/or degree of thiol substitution. The GelNB/HepSH hydrogel allowed little intracellular ROS for encapsulated cells but provided vascular endothelial growth factor binding affinity for potential facilitation of neovascularization. Finally, the GelNB/HepSH bioink enabled a convenient printing process for both complex-structured bioscaffolds and cell-laden constructs, and resulted in good printability and high post-crosslinking cell viability. The crosslinker HepSH may serve as a multifunctional macromolecule that enables GelNB-based bioprinting in broad applications in regenerative medicine.
Sigma-Aldrich
TissueFab® bioink , (GelHep)MA Vis/ 405nm, low endotoxin
Sigma-Aldrich
TissueFab® bioink , (GelHAHep)MA Vis/405 nm, low endotoxin