Merck
Endothelin-1 gene regulation.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2010-09-15)
Lisa R Stow, Mollie E Jacobs, Charles S Wingo, Brian D Cain
ABSTRACT
Over two decades of research have demonstrated that the peptide hormone endothelin-1 (ET-1) plays multiple, complex roles in cardiovascular, neural, pulmonary, reproductive, and renal physiology. Differential and tissue-specific production of ET-1 must be tightly regulated in order to preserve these biologically diverse actions. The primary mechanism thought to control ET-1 bioavailability is the rate of transcription from the ET-1 gene (edn1). Studies conducted on a variety of cell types have identified key transcription factors that govern edn1 expression. With few exceptions, the cis-acting elements bound by these factors have been mapped in the edn1 regulatory region. Recent evidence has revealed new roles for some factors originally believed to regulate edn1 in a tissue or hormone-specific manner. In addition, other mechanisms involved in epigenetic regulation and mRNA stability have emerged as important processes for regulated edn1 expression. The goal of this review is to provide a comprehensive overview of the specific factors and signaling systems that govern edn1 activity at the molecular level.
Sigma-Aldrich
Endothelin 1, Human and Porcine, Endothelin 1, Human and Porcine, CAS 117399-94-7, is a 21-amino acid polypeptide with potent vasoconstrictive action. Activates phospholipase C in fibroblasts expressing ETA receptors.
Pricing and availability is not currently available.