Merck
  • Characterization of the humanized FRG mouse model and development of an AAV-LK03 variant with improved liver lobular biodistribution.
Characterization of the humanized FRG mouse model and development of an AAV-LK03 variant with improved liver lobular biodistribution.
Molecular therapy. Methods & clinical development (2023-01-27)
Marti Cabanes-Creus, Renina Gale Navarro, Sophia H Y Liao, Suzanne Scott, Rodrigo Carlessi, Ramon Roca-Pinilla, Maddison Knight, Grober Baltazar, Erhua Zhu, Matthew Jones, Elena Denisenko, Alistair R R Forrest, Ian E Alexander, Janina E E Tirnitz-Parker, Leszek Lisowski
ABSTRACT
Recent clinical successes have intensified interest in using adeno-associated virus (AAV) vectors for therapeutic gene delivery. The liver is a key clinical target, given its critical physiological functions and involvement in a wide range of genetic diseases. In the present study, we first investigated the validity of a liver xenograft mouse model repopulated with primary hepatocytes using single-nucleus RNA sequencing (sn-RNA-seq) by studying the transcriptomic profile of human hepatocytes pre- and post-engraftment. Complementary immunofluorescence analyses performed in highly engrafted animals confirmed that the human hepatocytes organize and present appropriate patterns of zone-dependent enzyme expression in this model. Next, we tested a set of rationally designed HSPG de-targeted AAV-LK03 variants for relative transduction performance in human hepatocytes. We used immunofluorescence, next-generation sequencing, and single-nucleus transcriptomics data from highly engrafted FRG mice to demonstrate that the optimally HSPG de-targeted AAV-LK03 displayed a significantly improved lobular transduction profile in this model.
Sigma-Aldrich
Goat Anti-Human IgG Antibody, HRP conjugate, 1.0 mg/mL, Chemicon®
Sigma-Aldrich
Anti-HAL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution