Merck
  • Optimization of slow cooling cryopreservation for human pluripotent stem cells.
Optimization of slow cooling cryopreservation for human pluripotent stem cells.
Genesis (New York, N.Y. : 2000) (2013-11-21)
Takamichi Miyazaki, Norio Nakatsuji, Hirofumi Suemori
ABSTRACT
Human pluripotent stem cells (hPSCs) have the potential for unlimited expansion and differentiation into cell types of all three germ layers. Cryopreservation is a key process for successful application of hPSCs. However, the current conventional method leads to poor recovery of hPSCs after thawing. Here, we demonstrate a highly efficient recovery method for hPSC cryopreservation by slow freezing and single-cell dissociation. After confirming hPSC survivability after freeze-thawing, we found that hPSCs that were freeze-thawed as colonies showed markedly decreased survival, whereas freeze-thawed single hPSCs retained the majority of their viability. These observations indicated that hPSCs should be cryopreserved as single cells. Freeze-thawed single hPSCs efficiently adhered and survived in the absence of a ROCK inhibitor by optimization of the seeding density. The high recovery rate enabled conventional colony passaging for subculture within 3 days post-thawing. The improved method was also adapted to a xeno-free culture system. Moreover, the cell recovery postcryopreservation was highly supported by coating culture surfaces with human laminin-521 that promotes adhesion of dissociated single hPSCs. This simplified but highly efficient cryopreservation method allows easy handling of cells and bulk storage of high-quality hPSCs.
Sigma-Aldrich
Anti-TRA-1-60 Antibody, clone TRA-1-60, clone TRA-1-60, Chemicon®, from mouse
Sigma-Aldrich
Anti-HLA-DRB1 Antibody, clone DJ9, clone DJ9, from mouse