Merck
  • SWI/SNF Antagonism of PRC2 Mediates Estrogen-Induced Progesterone Receptor Expression.
SWI/SNF Antagonism of PRC2 Mediates Estrogen-Induced Progesterone Receptor Expression.
Cells (2022-03-26)
Mike R Wilson, Jake J Reske, Julie Koeman, Marie Adams, Niraj R Joshi, Asgerally T Fazleabas, Ronald L Chandler
ABSTRACT
Endometrial cancer (EC) is characterized by high estrogen levels unopposed by progesterone. Treatment with progestins is standard for early EC, but the response to progestins is dependent on progesterone receptor (PGR) expression. Here, we show that the expression of PGR in endometrial epithelial cells is dependent on ARID1A, a DNA-binding subunit of the SWI/SNF chromatin-remodeling complex that is commonly mutated in EC. In endometrial epithelial cells with estrogen receptor overexpression, we find that ARID1A promotes estrogen signaling and regulates common gene expression programs. Normally, endometrial epithelial cells expressing estrogen receptors respond to estrogen by upregulating the PGR. However, when ARID1A expression is lost, upregulation of PGR expression is significantly reduced. This phenomenon can also occur following the loss of the SWI/SNF subunit BRG1, suggesting a role for ARID1A- and BRG1-containing complexes in PGR regulation. We find that PGR is regulated by a bivalent promoter, which harbors both H3K4me3 and H3K27me3 histone tail modifications. H3K27me3 is deposited by EZH2, and inhibition of EZH2 in the context of ARID1A loss results in restoration of estrogen-induced PGR expression. Our results suggest a role for ARID1A deficiency in the loss of PGR in late-stage EC and a therapeutic utility for EZH2 inhibitors in this disease.
Sigma-Aldrich
Anti-Progesterone Receptor antibody, Rabbit monoclonal, recombinant, expressed in proprietary host, clone SP2, tissue culture supernatant