Merck
  • Crystal structure of the full Swi2/Snf2 remodeler Mot1 in the resting state.
Crystal structure of the full Swi2/Snf2 remodeler Mot1 in the resting state.
eLife (2018-10-06)
Agata Butryn, Stephan Woike, Savera J Shetty, David T Auble, Karl-Peter Hopfner
ABSTRACT
Swi2/Snf2 ATPases remodel protein:DNA complexes in all of the fundamental chromosome-associated processes. The single-subunit remodeler Mot1 dissociates TATA box-binding protein (TBP):DNA complexes and provides a simple model for obtaining structural insights into the action of Swi2/Snf2 ATPases. Previously we reported how the N-terminal domain of Mot1 binds TBP, NC2 and DNA, but the location of the C-terminal ATPase domain remained unclear (Butryn et al., 2015). Here, we report the crystal structure of the near full-length Mot1 from Chaetomium thermophilum. Our data show that Mot1 adopts a ring like structure with a catalytically inactive resting state of the ATPase. Biochemical analysis suggests that TBP binding switches Mot1 into an ATP hydrolysis-competent conformation. Combined with our previous results, these data significantly improve the structural model for the complete Mot1:TBP:DNA complex and suggest a general mechanism for Mot1 action.
Sigma-Aldrich
B834(DE3) Competent Cells - Novagen, B834 is the parental strain for BL21. These hosts are methionine auxotrophs and allow high specific activity labeling of target proteins with 35S-methionine and selenomethionine for crystallography.
Sigma-Aldrich
Rosetta(DE3) Competent Cells - Novagen, Rosetta host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli.