Merck
Green synthesis of size controllable gold nanoparticles.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-08-27)
Kesarla Mohan Kumar, Badal Kumar Mandal, Hoskote A Kiran Kumar, Sireesh Babu Maddinedi
ABSTRACT
A facile rapid green eco-friendly method to synthesize gold nanoparticles (Au NPs) of tunable size using aqueous Terminalia arjuna fruit extracts has been demonstrated herein. Formation of Au NPs was confirmed by Surface Plasmon Resonance (SPR) study at 528 nm using UV-visible spectrophotometer. The time of reduction, size and morphological variations of Au NPs were studied with varying quantities of T. arjuna fruit aqueous extracts. Synthesized Au NPs were characterized using UV-visible spectroscopy, Fourier transformed infrared spectroscopy (FT-IR), powder X-ray diffraction (XRD), transmission electron microscopy (TEM) and Energy dispersive X-ray spectroscopy (EDAX). Polyphenols responsible for reduction of Au(3+) to Au(0) were identified using High Performance Liquid Chromatography (HPLC) as ascorbic acid, gallic acid and pyrogallol. The oxidized forms of polyphenols formed coordination with surface of Au NPs which protected their further growth and aggregation. We also propose a plausible mechanism how to tune size and shape of Au NPs by varying the quantity of extracts. Thus obtained Au NPs were stable for more than four months.
Sigma-Aldrich
Diastase from Aspergillus oryzae, powder, white, amylase ≥3500 U/g
Pricing and availability is not currently available.