Merck
Anti-tumor effects of crocetin and related molecular targets.
Journal of cellular physiology (2017-04-14)
Maliheh Moradzadeh, Hamid Reza Sadeghnia, Alijan Tabarraei, Amirhossein Sahebkar
ABSTRACT
Natural products have gained a wide popularity as chemopreventive and anti-cancer agents owing to their multi-mechanistic mode of action, availability and synergism with several conventional chemotherapeutic agents. Crocetin is a carotenoid compound isolated from the stigma of Crocus sativus L. (saffron). Crocetin has shown promising effects as an anti-tumor agent in animal models and cell culture systems. Crocetin retards the growth of cancer cells via inhibiting nucleic acid synthesis, enhancing anti-oxidative system, and inducing apoptosis and differentiation pathways. The present review outlines natural sources of crocetin, and its pharmacokinetic and pharmacological properties relevant to the prevention and treatment of cancer. Also, we discuss molecular targets underlying the putative anti-tumor effects of crocetin.
Sigma-Aldrich
Crocetin, ≥98% (HPLC)