Merck
  • Protocol for accelerated skeletal muscle regeneration and hypertrophic muscle formation in mice.
Protocol for accelerated skeletal muscle regeneration and hypertrophic muscle formation in mice.
STAR protocols (2022-02-05)
Rashmi Ray, Vivek Rai
ABSTRACT
The skeletal muscle system is the major organ associated with movement of the body. Myogenesis and regeneration induced post-injury contribute to muscle formation and maintenance. Here, we provide detailed protocol for the accelerated repair of injured skeletal muscles and generation of hypertrophic muscle fibers. This protocol includes cardiotoxin induced muscle injury and also describes isolation of satellite cells from skeletal muscle tissues of mice. This protocol can be used to study the mechanisms associated with accelerated muscle repair and hypertrophy. For complete details on the use and execution of this protocol, please refer to Ray et al. (2021).
Sigma-Aldrich
Collagenase from Clostridium histolyticum, non-sterile; 0.2 μm filtered, Type IA-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture