Merck
  • Oxidative markers, nitric oxide and homocysteine alteration in hypercholesterolimic rats: role of atorvastatine and cinnamon.
Oxidative markers, nitric oxide and homocysteine alteration in hypercholesterolimic rats: role of atorvastatine and cinnamon.
International journal of clinical and experimental medicine (2009-11-18)
Kamal A Amin, Thanaa M Abd El-Twab
ABSTRACT
To investigate the effects of atorvastatin and cinnamon on serum lipid profile, oxidative stress, antioxidant capacity, hepatic enzymes activities, nitric oxide (NO) as well as homocysteine (Hcy) in hypercholesterolemic rats, 48 male albino rats, weighing 130-190 gm were divided into 2 groups, normal group fed on basal rat chow diet (n=12) and high cholesterol group (HCD) were fed on 1% cholesterol-enriched diet for 15 day (n=36). Hypercholesterolemic rats were divided into 3 subgroups (n=12 for each) fed the same diet and treated with atorvastatine (HCD+Atorvastatin) or cinnamon extract (HCD+cinnamon) or none treated (HCD) for 3&6 weeks. Serum triglycerides (TG), Total cholesterol (TC), low density lipoprotein (LDL), high density lipoprotein (HDL), ALT, AST, NO, Hcy, hepatic reduced glutathione (GSH), Malondialdehyde (MDA) and antioxidant enzymes, Superoxide dismutase (SOD) and catalase activity were measured. Results showed that HCD increased significantly TG, TC, LDL-C, ALT, AST, Hcy and hepatic MDA, while lowered significantly antioxidant enzyme activities and NO levels. Atorvastatin therapy significantly increased HDL-C, NO and antioxidant activity while decreased LDL-C, MDA and Hcy concentrations. Serum TG, TC, LDL-C, ALT, AST and hepatic MDA levels were significantly lowered meanwhile, serum HDL, NO values and hepatic antioxidant activities were significantly, higher in cinnamon-treated than untreated group. These results indicate that lipid abnormalities, oxidative injury and hyperhomocystienemia were induced by HCD and this study recommend that administration of atorvastatine or cinnamon provided protection against the lipemic-oxidative disorder and act as hypocholesterolemic, hepatoprotective agent and improve cardiovascular function through modulation of oxidative stress, NO and Hcy.
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%