Merck
  • Engineering glycoside hydrolase stability by the introduction of zinc binding.
Engineering glycoside hydrolase stability by the introduction of zinc binding.
Acta crystallographica. Section D, Structural biology (2018-07-04)
Thomas L Ellinghaus, Jose H Pereira, Ryan P McAndrew, Ditte H Welner, Andy M DeGiovanni, Joel M Guenther, Huu M Tran, Taya Feldman, Blake A Simmons, Kenneth L Sale, Paul D Adams
ABSTRACT
The development of robust enzymes, in particular cellulases, is a key step in the success of biological routes to `second-generation' biofuels. The typical sources of the enzymes used to degrade biomass include mesophilic and thermophilic organisms. The endoglucanase J30 from glycoside hydrolase family 9 was originally identified through metagenomic analyses of compost-derived bacterial consortia. These studies, which were tailored to favor growth on targeted feedstocks, have already been shown to identify cellulases with considerable thermal tolerance. The amino-acid sequence of J30 shows comparably low identity to those of previously analyzed enzymes. As an enzyme that combines a well measurable activity with a relatively low optimal temperature (50°C) and a modest thermal tolerance, it offers the potential for structural optimization aimed at increased stability. Here, the crystal structure of wild-type J30 is presented along with that of a designed triple-mutant variant with improved characteristics for industrial applications. Through the introduction of a structural Zn2+ site, the thermal tolerance was increased by more than 10°C and was paralleled by an increase in the catalytic optimum temperature by more than 5°C.
Sigma-Aldrich
4-Nitrophenyl β-D-cellobioside, ≥98% (TLC)