Merck
  • Manipulation of TAD reorganization by chemical-dependent genome linking.
Manipulation of TAD reorganization by chemical-dependent genome linking.
STAR protocols (2021-09-17)
Jia Wang, Qian Ma, Peihang Fang, Qi Tian, Haopeng Yu, Junyi Sun, Junjun Ding
ABSTRACT
Reorganization of topologically associated domain (TAD) is considered to be a novel mechanism for cell fate transitions. Here, we present a protocol to manipulate TAD via abscisic acid (ABA)-dependent genome linking. We use this protocol to merge two adjacent TADs and evaluate the influence on cell fate transitions. The advantages are that the manipulation does not change the genome and is reversible by withdrawing ABA. The major challenge is how to select linking loci for efficient TAD reorganization. For complete details on the use and execution of this protocol, please refer to Wang et al. (2021).
Sigma-Aldrich
Anti-CTCF Antibody, serum, Upstate®
Sigma-Aldrich
Leukocyte Alkaline Phosphatase Kit, based on naphthol AS-BI and fast red violet LB
Sigma-Aldrich
Normal Rabbit IgG, Normal Rabbit IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
Sigma-Aldrich
Normal Mouse IgG, Normal Mouse IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.