Merck
  • Disrupting biological sensors of force promotes tissue regeneration in large organisms.
Disrupting biological sensors of force promotes tissue regeneration in large organisms.
Nature communications (2021-09-08)
Kellen Chen, Sun Hyung Kwon, Dominic Henn, Britta A Kuehlmann, Ruth Tevlin, Clark A Bonham, Michelle Griffin, Artem A Trotsyuk, Mimi R Borrelli, Chikage Noishiki, Jagannath Padmanabhan, Janos A Barrera, Zeshaan N Maan, Teruyuki Dohi, Chyna J Mays, Autumn H Greco, Dharshan Sivaraj, John Q Lin, Tobias Fehlmann, Alana M Mermin-Bunnell, Smiti Mittal, Michael S Hu, Alsu I Zamaleeva, Andreas Keller, Jayakumar Rajadas, Michael T Longaker, Michael Januszyk, Geoffrey C Gurtner
ABSTRACT
Tissue repair and healing remain among the most complicated processes that occur during postnatal life. Humans and other large organisms heal by forming fibrotic scar tissue with diminished function, while smaller organisms respond with scarless tissue regeneration and functional restoration. Well-established scaling principles reveal that organism size exponentially correlates with peak tissue forces during movement, and evolutionary responses have compensated by strengthening organ-level mechanical properties. How these adaptations may affect tissue injury has not been previously examined in large animals and humans. Here, we show that blocking mechanotransduction signaling through the focal adhesion kinase pathway in large animals significantly accelerates wound healing and enhances regeneration of skin with secondary structures such as hair follicles. In human cells, we demonstrate that mechanical forces shift fibroblasts toward pro-fibrotic phenotypes driven by ERK-YAP activation, leading to myofibroblast differentiation and excessive collagen production. Disruption of mechanical signaling specifically abrogates these responses and instead promotes regenerative fibroblast clusters characterized by AKT-EGR1.
Sigma-Aldrich
Anti-MFGE8 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution