Merck
  • Bio-Guided Isolation of Two New Hypoglycemic Triterpenoid Saponins from Polygonum capitatum.
Bio-Guided Isolation of Two New Hypoglycemic Triterpenoid Saponins from Polygonum capitatum.
Drug design, development and therapy (2021-12-25)
Doudou Huang, Zenan Du, Yanhong Chen, Zhiying Dong, Xiujuan Wang, Mengshuang Li, Feng Zhang, Wansheng Chen, Lianna Sun
ABSTRACT
Diabetes is a common disease caused by a combination of genetic and environmental factors, which was the top three diseases threatening human health. Therefore, it is necessary to seek more efficient hypoglycemic drugs. The main objective of this study was to investigate the potential hypoglycemic effects of compounds from Polygonum capitatum. Our experiments were divided into three steps: (1) α-amylase test and oral starch tolerance test (OSTT) for screening the biological extract part of P. capitatum; (2) chemical isolation and identification using various separation techniques, and spectrum methods; and (3) evaluation of α-amylase inhibitory activity of isolates and in silico analysis for mechanism investigation. The n-butanol fractioned part of P. capitatum was confirmed to be the biological part according to α-amylase test. Then, two new triterpenoid saponins were isolated from the n-butanol part, which were also the first isolated triterpenoid saponins from P. capitatum. The activities of compounds 1 and 2 against α-amylase were 51.9±2.8% and 38.1±2.2%, respectively, which was consistent with the molecular docking analysis. In which, 1 and 2 showed the binding affinity energy for α-amylase was -9.4 kcal/mol and -7.8 kcal/mol, respectively. Two new triterpenoid saponins were firstly isolated from P. capitatum, and displays potency as a hypoglycemic agent through blocking α-amylase.
Sigma-Aldrich
α-Amylase from porcine pancreas, Type I-A, PMSF treated, saline suspension, 700-1400 units/mg protein (E1%/280)