Merck
  • Proteomics-Based Identification of DUB Substrates Using Selective Inhibitors.
Proteomics-Based Identification of DUB Substrates Using Selective Inhibitors.
Cell chemical biology (2020-10-03)
Jonathan W Bushman, Katherine A Donovan, Nathan J Schauer, Xiaoxi Liu, Wanyi Hu, Anthony C Varca, Sara J Buhrlage, Eric S Fischer
ABSTRACT
Deubiquitinating enzymes (DUBs) catalyze the removal of ubiquitin, thereby reversing the activity of E3 ubiquitin ligases and are central to the control of protein abundance and function. Despite the growing interest in DUBs as therapeutic targets, cellular functions for DUBs remain largely unknown and technical challenges often preclude the identification of DUB substrates in a comprehensive manner. Here, we demonstrate that treatment with potent DUB inhibitors coupled to mass spectrometry-based proteomics can identify DUB substrates at a proteome-wide scale. We applied this approach to USP7, a DUB widely investigated as a therapeutic target and identified many known substrates and additional targets. We demonstrate that USP7 substrates are enriched for DNA repair enzymes and E3 ubiquitin ligases. This work provides not only a comprehensive annotation of USP7 substrates, but a general protocol widely applicable to other DUBs, which is critical for translational development of DUB targeted agents.
Product Description
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial
Sigma-Aldrich
XL188, ≥98% (HPLC)
Sigma-Aldrich
XL177B, ≥98% (HPLC)
Sigma-Aldrich
XL177A, ≥98% (HPLC)