Merck
  • ROS and Oxidative Stress Are Elevated in Mitosis during Asynchronous Cell Cycle Progression and Are Exacerbated by Mitotic Arrest.
ROS and Oxidative Stress Are Elevated in Mitosis during Asynchronous Cell Cycle Progression and Are Exacerbated by Mitotic Arrest.
Cell systems (2019-02-25)
Jesse C Patterson, Brian A Joughin, Bert van de Kooij, Daniel C Lim, Douglas A Lauffenburger, Michael B Yaffe
ABSTRACT
Although elevated levels of reactive oxygen species (ROS) have been observed in cancer cells and cancer cells aberrantly proliferate, it is not known whether the level of reactive oxygen species and the accumulation of oxidative damage to macromolecules vary across the cell cycle. Here, we measure the prevalence of reactive oxygen species and of biomolecule oxidation across the cell cycle in freely cycling cancer cells. We report that reactive oxygen species vary during the cell cycle and peak in mitosis, resulting in mitotic accumulation of oxidized protein cysteine residues. Prolonged mitotic arrest further increased the levels of ROS and the abundance of oxidatively damaged biomolecules, including cysteine-sulfenic-acid-containing proteins and 8-oxoguanine. These finding suggest that mitotic arrest agents may enhance the effects of ROS-dependent anticancer therapies.
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Supelco
5,5-Dimethyl-1,3-cyclohexanedione, for HPLC derivatization, for the determination of aldehyde formaldehyde, ≥99.0%
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, clone 3H10, clone 3H10, Upstate®, from mouse
Sigma-Aldrich
Anti-8-Oxoguanine Antibody, clone 483.15, ascites fluid, clone 483.15, Chemicon®