Merck
  • Sterols lower energetic barriers of membrane bending and fission necessary for efficient clathrin-mediated endocytosis.
Sterols lower energetic barriers of membrane bending and fission necessary for efficient clathrin-mediated endocytosis.
Cell reports (2021-11-18)
Ruthellen H Anderson, Kem A Sochacki, Harika Vuppula, Brandon L Scott, Elizabeth M Bailey, Maycie M Schultz, Jason G Kerkvliet, Justin W Taraska, Adam D Hoppe, Kevin R Francis
ABSTRACT
Clathrin-mediated endocytosis (CME) is critical for cellular signal transduction, receptor recycling, and membrane homeostasis in mammalian cells. Acute depletion of cholesterol disrupts CME, motivating analysis of CME dynamics in the context of human disorders of cholesterol metabolism. We report that inhibition of post-squalene cholesterol biosynthesis impairs CME. Imaging of membrane bending dynamics and the CME pit ultrastructure reveals prolonged clathrin pit lifetimes and shallow clathrin-coated structures, suggesting progressive impairment of curvature generation correlates with diminishing sterol abundance. Sterol structural requirements for efficient CME include 3' polar head group and B-ring conformation, resembling the sterol structural prerequisites for tight lipid packing and polarity. Furthermore, Smith-Lemli-Opitz fibroblasts with low cholesterol abundance exhibit deficits in CME-mediated transferrin internalization. We conclude that sterols lower the energetic costs of membrane bending during pit formation and vesicular scission during CME and suggest that reduced CME activity may contribute to cellular phenotypes observed within disorders of cholesterol metabolism.
Sigma-Aldrich
Latrunculin B, Latrunculia magnifica, Latrunculin B, CAS 76343-94-7, is a unique marine toxin that inhibits actin polymerization and disrupts microfilament organization. It is 10 to 100-fold more potent than cytochalasins.
Sigma-Aldrich
Cholesterol-Water Soluble, powder, BioReagent, suitable for cell culture