Merck
Supramolecular Atropine Potentiometric Sensor.
Sensors (Basel, Switzerland) (2021-09-11)
Catarina Ferreira, Andreia Palmeira, Emília Sousa, Célia G Amorim, Alberto Nova Araújo, Maria Conceição Montenegro
ABSTRACT
A supramolecular atropine sensor was developed, using cucurbit[6]uril as the recognition element. The solid-contact electrode is based on a polymeric membrane incorporating cucurbit[6]uril (CB[6]) as an ionophore, 2-nitrophenyl octyl ether as a solvent mediator, and potassium tetrakis (4-chlorophenyl) borate as an additive. In a MES-NaOH buffer at pH 6, the performance of the atropine sensor is characterized by a slope of (58.7 ± 0.6) mV/dec with a practical detection limit of (6.30 ± 1.62) × 10-7 mol/L and a lower limit of the linear range of (1.52 ± 0.64) × 10-6 mol/L. Selectivity coefficients were determined for different ions and excipients. The obtained results were bolstered by the docking and spectroscopic studies which demonstrated the interaction between atropine and CB[6]. The accuracy of the potentiometric analysis of atropine content in certified reference material was evaluated by the t-Student test. The herein proposed sensor answers the need for reliable methods providing better management of this hospital drug shelf-life while reducing its flush and remediation costs.
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Trizma® hydrochloride, reagent grade, ≥99.0% (titration), crystalline