Merck
  • Synthesis and Bioconjugation of Thiol-Reactive Reagents for the Creation of Site-Selectively Modified Immunoconjugates.
Synthesis and Bioconjugation of Thiol-Reactive Reagents for the Creation of Site-Selectively Modified Immunoconjugates.
Journal of visualized experiments : JoVE (2019-03-26)
Maria Davydova, Guillaume Dewaele Le Roi, Pierre Adumeau, Brian M Zeglis
ABSTRACT
Maleimide-bearing bifunctional probes have been employed for decades for the site-selective modification of thiols in biomolecules, especially antibodies. Yet maleimide-based conjugates display limited stability in vivo because the succinimidyl thioether linkage can undergo a retro-Michael reaction. This, of course, can lead to the release of the radioactive payload or its exchange with thiol-bearing biomolecules in circulation. Both of these processes can produce elevated activity concentrations in healthy organs as well as decreased activity concentrations in target tissues, resulting in reduced imaging contrast and lower therapeutic ratios. In 2018, we reported the creation of a modular, stable, and easily accessible phenyloxadiazolyl methyl sulfone reagent - dubbed 'PODS' - as a platform for thiol-based bioconjugations. We have clearly demonstrated that PODS-based site-selective bioconjugations reproducibly and robustly create homogenous, well-defined, highly immunoreactive, and highly stable radioimmunoconjugates. Furthermore, preclinical experiments in murine models of colorectal cancer have shown that these site-selectively labeled radioimmunoconjugates exhibit far superior in vivo performance compared to radiolabeled antibodies synthesized via maleimide-based conjugations. In this protocol, we will describe the four-step synthesis of PODS, the creation of a bifunctional PODS-bearing variant of the ubiquitous chelator DOTA (PODS-DOTA), and the conjugation of PODS-DOTA to the HER2-targeting antibody trastuzumab.
Sigma-Aldrich
Phosphate buffered saline, 10× concentrate, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
N-Boc-N′-succinyl-4,7,10-trioxa-1,13-tridecanediamine, 95% (HPLC)