Merck
  • Fabrication of alginate-based hydrogel cross-linked via horseradish peroxidase for articular cartilage engineering.
Fabrication of alginate-based hydrogel cross-linked via horseradish peroxidase for articular cartilage engineering.
BMC research notes (2021-09-30)
Sepideh Saghati, Ali Baradar Khoshfetrat, Hamid Tayefi Nasrabadi, Leila Roshangar, Reza Rahbarghazi
ABSTRACT
We aimed to detect the effect of a couple of parameters including Alg, H2O2, and HRP concentrations on the gelation time of Alg-based hydrogels using an enzymatic cross-linked procedure. NMR, UV-Vis, and ATR-FTIR analyses confirmed the conjugation of Ph to the Alg backbone. Data showed gelation time was delayed with the increase and reduction of H2O2 and HRP, respectively. We noted that hydrogel consisted of 1.2% (w/v) Alg, 5 U HRP, and 100 mM H2O2 yielded an appropriate gelation time with appropriate mechanical properties. The addition of 0.5% (v/v) Col developed hydrogel increased the gelation time. The data showed that Alg, HRP, and H2O2 with the ratio of 1:0.54:0.54 had proper physicochemical features for cartilage engineering.
Sigma-Aldrich
Tyramine hydrochloride, ≥98%
Pricing and availability is not currently available.