Merck
  • Amphotericin B induces epithelial voltage responses in people with cystic fibrosis.
Amphotericin B induces epithelial voltage responses in people with cystic fibrosis.
Journal of cystic fibrosis : official journal of the European Cystic Fibrosis Society (2020-12-15)
Rajeev S Chorghade, Bo Ram Kim, Janice L Launspach, Philip H Karp, Michael J Welsh, Martin D Burke
ABSTRACT
Approximately 10% of people with cystic fibrosis (CF) have mutations that result in little to no CFTR production and thus cannot benefit from CFTR modulators. We previously found that Amphotericin B (AmB), a small molecule that forms anion channels, restored HCO3- secretion and increased host defenses in primary cultures of CF airway epithelia. Further, AmB increased ASL pH in CFTR-null pigs, suggesting an alternative CFTR-independent approach to achieve gain-of-function. However, it remains unclear whether this approach can be effective in people. To determine whether AmB can impact physiology in people with CF, we first tested whether Fungizone, a clinically approved AmB formulation, could cause electrophysiological effects consistent with anion secretion in primary cultures of CF airway epithelia. We then evaluated the capacity of AmB to change nasal potential difference (NPD), a key clinical biomarker, in people with CF not on CFTR modulators. AmB increased transepithelial Cl- current and hyperpolarized calculated transepithelial voltage in primary cultures of CF airway epithelia from people with two nonsense mutations. In eight people with CF not on CFTR modulators, intranasal Fungizone treatment caused a statistically significant change in NPD. This change was similar in direction and magnitude to the effect of ivacaftor in people with a G551D mutation. Our results provide the first evidence that AmB can impact a clinical biomarker in people with CF. These results encourage additional clinical studies in people with CF to determine whether small molecule anion channels can provide benefit.
Sigma-Aldrich
Gentamicin solution, 50 mg/mL in deionized water, liquid, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
D-Glucose-1-13C, 99 atom % 13C
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Sodium bicarbonate-13C, 98 atom % 13C, 99% (CP)
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Amiloride hydrochloride hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Fluconazole, ≥98% (HPLC), powder