Merck
  • Therapeutic Potential of NTRK3 Inhibition in Desmoplastic Small Round Cell Tumor.
Therapeutic Potential of NTRK3 Inhibition in Desmoplastic Small Round Cell Tumor.
Clinical cancer research : an official journal of the American Association for Cancer Research (2020-11-25)
Koichi Ogura, Romel Somwar, Julija Hmeljak, Heather Magnan, Ryma Benayed, Amir Momeni Boroujeni, Anita S Bowman, Marissa S Mattar, Inna Khodos, Elisa de Stanchina, Achim Jungbluth, Marina Asher, Igor Odintsov, Alifiani B Hartono, Michael P LaQuaglia, Emily Slotkin, Christine A Pratilas, Sean Bong Lee, Lee Spraggon, Marc Ladanyi
ABSTRACT
Desmoplastic small round cell tumor (DSRCT) is a highly lethal intra-abdominal sarcoma of adolescents and young adults. DSRCT harbors a t(11;22)(p13:q12) that generates the EWSR1-WT1 chimeric transcription factor, the key oncogenic driver of DSRCT. EWSR1-WT1 rewires global gene expression networks and activates aberrant expression of targets that together mediate oncogenesis. EWSR1-WT1 also activates a neural gene expression program. Among these neural markers, we found prominent expression of neurotrophic tyrosine kinase receptor 3 (NTRK3), a druggable receptor tyrosine kinase. We investigated the regulation of NTRK3 by EWSR1-WT1 and its potential as a therapeutic target in vitro and in vivo, the latter using novel patient-derived models of DSRCT. We found that EWSR1-WT1 binds upstream of NTRK3 and activates its transcription. NTRK3 mRNA is highly expressed in DSRCT compared with other major chimeric transcription factor-driven sarcomas and most DSRCTs are strongly immunoreactive for NTRK3 protein. Remarkably, expression of NTRK3 kinase domain mRNA in DSRCT is also higher than in cancers with NTRK3 fusions. Abrogation of NTRK3 expression by RNAi silencing reduces growth of DSRCT cells and pharmacologic targeting of NTRK3 with entrectinib is effective in both in vitro and in vivo models of DSRCT. Our results indicate that EWSR1-WT1 directly activates NTRK3 expression in DSRCT cells, which are dependent on its expression and activity for growth. Pharmacologic inhibition of NTRK3 by entrectinib significantly reduces growth of DSRCT cells both in vitro and in vivo, providing a rationale for clinical evaluation of NTRK3 as a therapeutic target in DSRCT.
Product Description
Roche
cOmplete ULTRA Tablets, Mini, EDTA-free, EASYpack Protease Inhibitor Cocktail, Tablets supplied in foil blister packs.
Pricing and availability is not currently available.