Merck
  • On the importance of the linking chemistry for the PEGylation of mesoporous silica nanoparticles.
On the importance of the linking chemistry for the PEGylation of mesoporous silica nanoparticles.
Journal of colloid and interface science (2021-01-24)
Cornelia von Baeckmann, Hanspeter Kählig, Mika Lindén, Freddy Kleitz
ABSTRACT
The typical method for minimizing serum protein adsorption in biological settings and prolonging blood circulation time of nanoparticles, is to anchor hydrophilic polymers (e.g., poly(ethylene glycol), PEG) on the particle surface, which is most often done by covalent attachment (PEGylation). Herein, different PEGylation methods were realised and compared to functionalize mesoporous silica nanoparticles (MSNs). First, reactive groups were installed using post-grafting procedures with different functional silanes. Further, PEGs carrying a functional group and having different chain lengths and termini, were used. The grafting efficacy as well as the structural and physicochemical characteristics of the resulting particles were determined. Finally, the serum protein adsorption behaviour of these functionalized particles was investigated using thermogravimetric analysis. The type of selected coupling method was shown to strongly influence the grafting efficiency as well as the resulting protein adsorption. The results highlight the importance of the right choice of the linking chemistry when aiming at surface functionalization of nanoparticles.
Sigma-Aldrich
(3-Mercaptopropyl)trimethoxysilane, 95%
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Allyl methyl sulfone, 96%
Sigma-Aldrich
Glutaric acid, 99%
Sigma-Aldrich
Triethoxysilane, 95%