Merck
  • Mutational upregulation of a resistance-nodulation-cell division-type multidrug efflux pump, SdeAB, upon exposure to a biocide, cetylpyridinium chloride, and antibiotic resistance in Serratia marcescens.
Mutational upregulation of a resistance-nodulation-cell division-type multidrug efflux pump, SdeAB, upon exposure to a biocide, cetylpyridinium chloride, and antibiotic resistance in Serratia marcescens.
Antimicrobial agents and chemotherapy (2009-09-16)
Hideaki Maseda, Yumiko Hashida, Rumi Konaka, Akihiro Shirai, Hiroki Kourai
ABSTRACT
Serratia marcescens is an important opportunistic pathogen in hospitals, where quaternary ammonium compounds are often used for disinfection. The aim of this study is to elucidate the effect of a biocide on the emergence of biocide- and antibiotic-resistant mutants and to characterize the molecular mechanism of biocide resistance in Serratia marcescens. A quaternary ammonium compound-resistant strain, CRes01, was selected by exposing a wild-type strain of S. marcescens to cetylpyridinium chloride. The CRes01 cells exhibited 2- to 16-fold more resistance than the wild-type cells to biocides and antibiotics, including cetylpyridinium chloride, benzalkonium chloride, chlorhexidine gluconate, fluoroquinolones, tetracycline, and chloramphenicol, and showed increased susceptibilities to beta-lactam antibiotics and N-dodecylpyridinium iodide. Mutant cells accumulated lower levels of norfloxacin than the parent cells in an energized state but not in a de-energized state, suggesting that the strain produced a multidrug efflux pump(s). To verify this assumption, we knocked out a putative efflux pump gene, sdeAB, in CRes01 and found that the knockout restored susceptibility to most quaternary ammonium compounds and antibiotics, to which the CRes01 strain showed resistance. On the basis of these and other results, we concluded that S. marcescens gains resistance to both biocides and antibiotics by expressing the SdeAB efflux pump upon exposure to cetylpyridinium chloride.
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Carbenicillin, Ready Made Solution, 100 mg/mL in ethanol/water, 0.2 μm filtered
Supelco
Norfloxacin, VETRANAL®, analytical standard
Supelco
Ofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Nalidixic acid, ≥98%
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Supelco
Norfloxacin, analytical standard, ≥98% (TLC)