Merck
Mild clinical presentation in KLHL40-related nemaline myopathy (NEM 8).
Neuromuscular disorders : NMD (2016-08-17)
Andreea M Seferian, Edoardo Malfatti, Caroline Bosson, Laurent Pelletier, Jessica Taytard, Veronique Forin, Teresa Gidaro, Elena Gargaun, Pierre Carlier, Julien Fauré, Norma B Romero, John Rendu, Laurent Servais
ABSTRACT
Nemaline myopathies are clinically and genetically heterogeneous muscle diseases characterized by the presence of nemaline bodies (rods) in muscle fibers. Mutations in the KLHL40 (kelch-like family member 40) gene (NEM 8) are common cause of severe/lethal nemaline myopathy. We report an 8-year-old girl born to consanguineous Moroccan parents, who presented with hypotonia and poor sucking at birth, delayed motor development, and further mild difficulties in walking and fatigability. A muscle biopsy revealed the presence of nemaline bodies. KLHL40 gene Sanger sequencing disclosed a never before reported pathogenic homozygous mutation which resulted in absent KLHL40 protein expression in the muscle. This further expands the phenotypical spectrum of KLHL40 related nemaline myopathy.
Sigma-Aldrich
Anti-KLHL40 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution