Merck
  • Design considerations of an IL13Rα2 antibody-drug conjugate for diffuse intrinsic pontine glioma.
Design considerations of an IL13Rα2 antibody-drug conjugate for diffuse intrinsic pontine glioma.
Acta neuropathologica communications (2021-05-19)
Xiaolei Lian, Dina Kats, Samuel Rasmussen, Leah R Martin, Anju Karki, Charles Keller, Noah E Berlow
ABSTRACT
Diffuse intrinsic pontine glioma (DIPG), a rare pediatric brain tumor, afflicts approximately 350 new patients each year in the United States. DIPG is noted for its lethality, as fewer than 1% of patients survive to five years. Multiple clinical trials involving chemotherapy, radiotherapy, and/or targeted therapy have all failed to improve clinical outcomes. Recently, high-throughput sequencing of a cohort of DIPG samples identified potential therapeutic targets, including interleukin 13 receptor subunit alpha 2 (IL13Rα2) which was expressed in multiple tumor samples and comparably absent in normal brain tissue, identifying IL13Rα2 as a potential therapeutic target in DIPG. In this work, we investigated the role of IL13Rα2 signaling in progression and invasion of DIPG and viability of IL13Rα2 as a therapeutic target through the use of immunoconjugate agents. We discovered that IL13Rα2 stimulation via canonical ligands demonstrates minimal impact on both the cellular proliferation and cellular invasion of DIPG cells, suggesting IL13Rα2 signaling is non-essential for DIPG progression in vitro. However, exposure to an anti-IL13Rα2 antibody-drug conjugate demonstrated potent pharmacological response in DIPG cell models both in vitro and ex ovo in a manner strongly associated with IL13Rα2 expression, supporting the potential use of targeting IL13Rα2 as a DIPG therapy. However, the tested ADC was effective in most but not all cell models, thus selection of the optimal payload will be essential for clinical translation of an anti-IL13Rα2 ADC for DIPG.
Sigma-Aldrich
Monoclonal Anti-IL13RA2 antibody produced in mouse, clone 2E10, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Goat anti-GAPDH Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.