Merck
  • Carbonic anhydrase inhibitors: inhibition studies of a coral secretory isoform with inorganic anions.
Carbonic anhydrase inhibitors: inhibition studies of a coral secretory isoform with inorganic anions.
Bioorganic & medicinal chemistry letters (2009-01-06)
Anthony Bertucci, Alessio Innocenti, Didier Zoccola, Andrea Scozzafava, Denis Allemand, Sylvie Tambutté, Claudiu T Supuran
ABSTRACT
The inhibition of a coral carbonic anhydrase (CA, EC 4.2.1.1) has been investigated with a series of inorganic anions such as halogenides, pseudohalogenides, bicarbonate, carbonate, nitrate, nitrite, hydrogen sulfide, bisulfite, perchlorate, sulfate. The full-length scleractinian coral Stylophora pistillata CA, STPCA, has a significant catalytic activity for the physiological reaction of CO(2) hydration to bicarbonate, similarly to the ubiquitous human isoforms hCA I (cytosolic) and hCA VI (secreted). The best STPCA anion inhibitors were bromide, iodide, carbonate, and sulfamate, with inhibition constants of 9.0-10.0 microM.
Sigma-Aldrich
Phenylboronic acid, 95%
Sigma-Aldrich
Sulfamide, 99%