Merck
  • Long-term epilepsy-associated tumors: transcriptional signatures reflect clinical course.
Long-term epilepsy-associated tumors: transcriptional signatures reflect clinical course.
Scientific reports (2020-01-11)
Daniel Delev, Karam Daka, Sabrina Heynckes, Annette Gaebelein, Pamela Franco, Dietmar Pfeifer, Marco Prinz, Oliver Schnell, Horst Urbach, Irina Mader, Jürgen Beck, Alexander Grote, Albert J Becker, Dieter Henrik Heiland
ABSTRACT
Long-term epilepsy-associated tumors (LEATs) represent mostly benign brain tumors associated with drug-resistant epilepsy. The aim of the study was to investigate the specific transcriptional signatures of those tumors and characterize their underlying oncogenic drivers. A cluster analysis of 65 transcriptome profiles from three independent datasets resulted in four distinct transcriptional subgroups. The first subgroup revealed transcriptional activation of STAT3 and TGF-signaling pathways and contained predominantly dysembryoplastic neuroepithelial tumors (DNTs). The second subgroup was characterized by alterations in the MAPK-pathway and up-stream cascades including FGFR and EGFR-mediated signaling. This tumor cluster exclusively contained neoplasms with somatic BRAFV600E mutations and abundance of gangliogliomas (GGs) with a significantly higher recurrence rate (42%). This finding was validated by examining recurrent tumors from the local database exhibiting BRAFV600E in 90% of the cases. The third cluster included younger patients with neuropathologically diagnosed GGs and abundance of the NOTCH- and mTOR-signaling pathways. The transcript signature of the fourth cluster (including both DNTs and GGs) was related to impaired neural function. Our analysis suggests distinct oncological pathomechanisms in long-term epilepsy-associated tumors. Transcriptional activation of MAPK-pathway and BRAFV600E mutation are associated with an increased risk for tumor recurrence and malignant progression, therefore the treatment of these tumors should integrate both epileptological and oncological aspects.
Sigma-Aldrich
Anti-BRAF (V600E) antibody, Rabbit monoclonal, recombinant, expressed in HEK 293 cells, clone RM8, purified immunoglobulin
Pricing and availability is not currently available.