Merck
  • Combining FRET and optical tweezers to study RhoGTPases spatio-temporal dynamics upon local stimulation.
Combining FRET and optical tweezers to study RhoGTPases spatio-temporal dynamics upon local stimulation.
Journal of biological methods (2017-03-07)
Federico Iseppon, Luisa Mr Napolitano, Vincent Torre, Dan Cojoc
ABSTRACT
Local stimulation with optical tweezers has been used to mimic natural stimuli that occur in biological processes such as cell migration or differentiation. Carriers (beads and lipid vesicles) with sizes down to 30 nm can be manipulated with a high spatial and temporal resolution: they are positioned with a sub-micrometric precision on a specific cell compartment and the beginning of the stimulation can be triggered with millisecond precision. RhoGTPases are a Ras-related family of proteins that regulate many different functions including cell polarity, microtubule dynamics and membrane transport pathways. Here we combine local stimulation with FRET microscopy to study RhoGTPases spatial and temporal activation following guidance cue local stimulation. We used two different vectors for local delivery: silica micro-beads and micro-sized lipid vesicles. The experimental methods associated with neuronal growth cone local stimulation are discussed in detail, as well as the analysis methods. Here we present a protocol that enables to study neuronal growth cone cytoskeleton rearrangements in response to a gradient of molecules in a way that better mimics physiological conditions, and it can be similarly applied to each secreted molecule involved in cell signaling.
Sigma-Aldrich
Liposome Kit: Lipid mixtures for the preparation of liposomes, lyophilized powder
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Bicinchoninic Acid Kit for Protein Determination, for 200-1000 μg/ml protein