Merck
  • Effect of biomolecules from human renal matrix of calcium oxalate monohydrate (CaOx) stones on in vitro calcium phosphate crystallization.
Effect of biomolecules from human renal matrix of calcium oxalate monohydrate (CaOx) stones on in vitro calcium phosphate crystallization.
International braz j urol : official journal of the Brazilian Society of Urology (2010-11-04)
Priyadarshini Pathak, Shrawan K Singh, Chanderdeep Tandon
ABSTRACT
Investigate the activity of high and low molecular weight biomolecules present in the matrix of human calcium oxalate (CaOx) stones not only on the initial mineral phase formation of calcium and phosphate (CaP) but also on its growth and demineralization of the preformed mineral phase. Surgically removed renal stones were analyzed by Fourier Transform Infra Red (FTIR) spectroscopy and only CaOx stones were extracted with 0.05M EGTA, 1 mM PMSF and 1% Β-mercaptoethanol. Renal CaOx stone extract was separated into > 10 kDa and < 10 kDa fractions by dialysis. Activity of both the fractions along with whole extract was studied on the three mineral phases of CaP assay system. It was interesting to observe that both high and low molecular weight biomolecules extracted from human renal matrix of calcium oxalate (CaOx) stones exhibited different roles in the three mineral phases of CaP. Whole extract exhibited inhibitory activity in all the three assay systems; however, mixed (stimulatory and inhibitory) activity was exhibited by the > 10 kDa and < 10 kDa fractions. SDS-PAGE analysis showed bands of 66 kDa, 80 kDa, 42 kDa in whole EGTA extract lane and > 10 kDa fraction lane. Both high and low molecular weight biomolecules extracted from human renal matrix of calcium oxalate (CaOx) stones have a significant influence on calcium and phosphate (CaP) crystallization.
Sigma-Aldrich
ProteoSilver Plus Silver Stain Kit