Merck
  • Synthesis and biological properties of 2-methylene-19-nor-25-dehydro-1alpha-hydroxyvitamin D(3)-26,23-lactones--weak agonists.
Synthesis and biological properties of 2-methylene-19-nor-25-dehydro-1alpha-hydroxyvitamin D(3)-26,23-lactones--weak agonists.
Bioorganic & medicinal chemistry (2008-08-30)
Grazia Chiellini, Pawel Grzywacz, Lori A Plum, Rafal Barycki, Margaret Clagett-Dame, Hector F DeLuca
ABSTRACT
In a continuing effort to explore the 2-methylene-1alpha-hydroxy-19-norvitamin D(3) class of pharmacologically important vitamin D compounds, two novel 2-methylene-19-nor-25-dehydro-1alpha-hydroxyvitamin D(3)-26,23-lactones, GC-3 and HLV, were synthesized and biologically tested. Based on reports of similarly structured molecules, it was hypothesized that these compounds might act as antagonists, at least in vitro. The pathway designed to synthesize these compounds was based on two key steps: first, the Lythgoe-type Wittig-Horner coupling of Windaus-Grundmann-type ketone 18, with phosphine oxide 15, followed, later in the synthesis, by the Zn-mediated Reformatsky-type allylation of aldehyde 20 with methylbromomethylacrylate 8. Our biological data show that neither compound has antagonistic activity but acts as weak agonists in vitro and in vivo.
Sigma-Aldrich
1α,25-Dihydroxyvitamin D2 solution, 50 μg/mL in ethanol, 95% (CP)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D2 solution, 100 μg/mL in ethanol, 95% (CP)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥99% (HPLC)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥97.0% (HPLC)