- Structural basis of reduction-dependent activation of human cystatin F.
Structural basis of reduction-dependent activation of human cystatin F.
Cystatins are important natural cysteine protease inhibitors targeting primarily papain-like cysteine proteases, including cathepsins and parasitic proteases like cruzipain, but also mammalian asparaginyl endopeptidase. Mammalian cystatin F, which is expressed almost exclusively in hematopoietic cells and accumulates in lysosome-like organelles, has been implicated in the regulation of antigen presentation and other immune processes. It is an unusual cystatin superfamily member with a redox-regulated activation mechanism and a restricted specificity profile. We describe the 2.1A crystal structure of human cystatin F in its dimeric "off" state. The two monomers interact in a fashion not seen before for cystatins or cystatin-like proteins that is crucially dependent on an unusual intermolecular disulfide bridge, suggesting how reduction leads to monomer formation and activation. Strikingly, core sugars for one of the two N-linked glycosylation sites of cystatin F are well ordered, and their conformation and interactions with the protein indicate that this unique feature of cystatin F may modulate its inhibitory properties, in particular its reduced affinity toward asparaginyl endopeptidase compared with other cystatins.