Merck
  • Induction of apoptosis in MCF-7:WS8 breast cancer cells by beta-lapachone.
Induction of apoptosis in MCF-7:WS8 breast cancer cells by beta-lapachone.
Cancer research (1998-05-15)
S M Wuerzberger, J J Pink, S M Planchon, K L Byers, W G Bornmann, D A Boothman
ABSTRACT
Beta-lapachone (beta-lap) affects a number of enzymes in vitro, including type I topoisomerase (Topo I); however, its exact intracellular target(s) and mechanism of cell killing remain unknown. We compared the cytotoxic responses of MCF-7:WS8 (MCF-7) human breast cancer cells after 4-h pulses of beta-lap or camptothecin (CPT), a known Topo I poison. A direct correlation between loss of survival and apoptosis was seen after beta-lap treatment (LD50 = 2.5 microM). A concentration-dependent, transient sub-2 N preapoptotic cell population was observed at 4-8 h. Estrogen deprivation-induced synchronization and bromodeoxyuridine-labeling studies revealed an apoptotic exit point near the G1-S border. Apoptosis activated by beta-lap was closely correlated with cleavage of lamin B but not with increases in p53/p21 or decreases in bcl-2. Loss of hyperphosphorylated forms of the retinoblastoma protein was observed within 5 h, but cyclins A, B1, and E levels were unaltered for up to 72 h after 5 microM beta-lap. Topo I and Topo IIalpha levels decreased at > 24 h. Logarithmic-phase MCF-7 cells were not affected by < or = 1 microM beta-lap. In contrast, dramatic and irreversible G2-M arrest with no apoptosis was observed in MCF-7 cells treated with 1 microM CPT, monitored for 6-10 days posttreatment. MCF-7 cells treated with supralethal doses of CPT (5 microM) resulted in only approximately 20% apoptosis. No correlation between apoptosis and loss of survival was observed. MCF-7 cells exposed to > 5 microM CPT arrested at key cell cycle checkpoints (i.e., G1, S, and G2-M), with little or no movement for 6 days. Ten-fold increases in p53/p21 and 2-5-fold decreases in bcl-2, Topo I, Topo IIalpha, and cyclins A and B1, with no change in cyclin E, were observed. Temporal decreases in bcl-2 and cleavage of lamin B corresponded to the minimal apoptotic response observed. Beta-lap activated apoptosis without inducing p53/p21 or cell cycle arrest responses and killed MCF-7 cells solely by apoptosis. In contrast, concentration-dependent increases in nuclear p53/p21 and various cell cycle checkpoint arrests were seen in MCF-7 cells after CPT. Despite dramatic p53/p21 protein induction responses, CPT-treated MCF-7 cells showed low levels of apoptosis, possibly due to protective cell cycle checkpoints or the lack of specific CPT-activated apoptotic pathways in MCF-7 cells.
Sigma-Aldrich
β-Lapachone, ≥98% (TLC)