Merck
  • Anti-inflammatory effects of ladostigil and its metabolites in aged rat brain and in microglial cells.
Anti-inflammatory effects of ladostigil and its metabolites in aged rat brain and in microglial cells.
Journal of neuroimmune pharmacology : the official journal of the Society on NeuroImmune Pharmacology (2012-03-29)
Rony Panarsky, Lisandro Luques, Marta Weinstock
ABSTRACT
Impaired mitochondrial function accompanied by microglial activation and the release of nitric oxide (NO) and pro-inflammatory cytokines has been reported in Alzheimer's disease, its prodromal phase of Mild Cognitive Impairment (MCI) and in aged rats. The present study showed that 6 months treatment of 16 month old rats with ladostigil (1 mg/kg/day), a novel drug designed for the treatment of MCI, prevented the development of spatial memory deficits at 22 months of age and significantly decreased the gene expression of IL-1β, IL-6, TNF-α and inducible nitric oxide synthase (iNOS) in the parietal cortex. It was also shown that concentrations ranging from 1nM-1 μM of ladostigil and three of its active metabolites inhibited the release of nitric oxide (NO) induced by lipopolysaccharide (LPS) from mouse microglial cells by up to 35-40 %. Ladostigil and its metabolites (10nM) also reduced TNF-α mRNA and protein by 25-35 % and IL-1β and inducible nitric oxide synthase (iNOS) mRNA by 20-35 %. The concentration of 10nM is in the range of that of the parent drug, R-MCPAI and R-HPAI found in plasma after oral administration of ladostigil (1 mg/kg/day) to rats. All the compounds inhibited the degradation of IkB-α and nuclear translocation of the p65 subunit of NF-kB. They also inhibited phosphorylation of p38 and ERK1/2 mitogen-activated protein kinase (MAPK), but had no effect on that of JNK. We propose that the anti-inflammatory activity may contribute towards the neuroprotective action of ladostigil against the development of memory impairments induced by aging or toxin-induced microglial activation.
Sigma-Aldrich
Ladostigil tartrate, ≥98% (HPLC)
Pricing and availability is not currently available.