Merck
  • Effect of mangiferin on hyperglycemia and atherogenicity in streptozotocin diabetic rats.
Effect of mangiferin on hyperglycemia and atherogenicity in streptozotocin diabetic rats.
Journal of ethnopharmacology (2005-03-03)
S Muruganandan, K Srinivasan, S Gupta, P K Gupta, J Lal
ABSTRACT
In the present study, the effect of mangiferin (a xanthone glucoside, isolated from the leaves of Mangifera indica) on the atherogenic potential of streptozotocin (STZ)-diabetes was investigated. In addition, the effect of mangiferin on oral glucose tolerance in glucose-loaded normal rats was also determined. The chronic intraperitoneal (i.p.) administration of mangiferin (10 and 20 mg/kg) once daily (o.d.) for 28 days exhibited antidiabetic activity by significantly lowering fasting plasma glucose level at different time intervals in STZ-diabetic rats. Further, mangiferin (10 and 20 mg/kg, i.p.) showed significant antihyperlipidemic and antiatherogenic activities as evidenced by significant decrease in plasma total cholesterol, triglycerides, low-density lipoprotein cholesterol (LDL-C) levels coupled together with elevation of high-density lipoprotein cholesterol (HDL-C) level and diminution of atherogenic index in diabetic rats. In addition, the chronic administration of mangiferin (10 and 20 mg/kg, i.p.) for 14 days significantly as well as markedly improved oral glucose tolerance in glucose-loaded normal rats suggesting its potent antihyperglycemic activity. The accumulating evidences suggest that both pancreatic and extrapancreatic mechanisms might be involved in its antidiabetic or antihyperglycemic action. In conclusion, the present study demonstrates that mangiferin possesses significant antidiabetic, antihyperlipidemic and antiatherogenic properties thus suggesting its beneficial effect in the treatment of diabetes mellitus associated with hyperlipidemia and related cardiovascular complications.
Sigma-Aldrich
Mangiferin Mangifera indica