Merck
  • The scabicide effect of moxidectin in vitro and in experimental animals: Parasitological, histopathological and immunological evaluation.
The scabicide effect of moxidectin in vitro and in experimental animals: Parasitological, histopathological and immunological evaluation.
Experimental parasitology (2020-08-11)
Mahmoud Sharaf, Sanaa Antonios, Samir Mina, Kamal Eliwa, Dina Abou Rayia
ABSTRACT
Scabies is considered one of the commonest dermatological diseases that has a global health burden. Current treatment with ivermectin (IVM) is insufficient and potential drug resistance was noticed. Moxidectin (MOX), with a better pharmacological profile may be a promising alternative. The efficacy of moxidectin against Sarcoptes scabiei was assessed both in vitro and in vivo in comparison with ivermectin. For the in vitro assay, both drugs were used in two concentrations (50 μg/ml and 100 μg/ml). For the in vivo assay, twenty rabbits infected with Sarcoptes scabiei were divided into three groups: untreated, moxidectin-treated and ivermectin-treated with the same dose of 0.3 mg/kg once. Another four rabbits were used as a normal control non-infected group. Treatment efficacy was evaluated by clinical assessment, parasitological evaluation and histopathological examination of skin samples using Hematoxylin and eosin and toluidine blue for mast cell staining. Immune response was also assessed by immunohistochemical staining of CD3 T cells in skin samples. Our results showed that moxidectin had a high efficacy (100%) in killing mites when used in both concentrations (50 μg/ml, 100 μg/ml) in the in vitro assay. Concerning the in vivo assay, on day 14 post-treatment, all MOX-treated rabbits were mite-free with full clinical cure by the end of the study (D21) showing (100%) reduction of mites count. Also, marked improvement in the epidermis with absence of mites in skin samples were shown. Poor clinical and parasitological improvements were noted in the ivermectin-treated rabbits, when given as a single dose with a percentage reduction (60.67%) in the 2nd week and progressive increase in lesions and mites count in the 3rd week post-treatment. Regarding the immune response, MOX-treated group showed mild infiltration with both mast cells and CD3 T cells in comparison to severe infiltration with both types of cells in the untreated and IVM-treated group. On conclusion, our results demonstrated that a single dose of MOX was more effective than IVM, supporting MOX as a valuable therapeutic approach for scabies therapy.
Sigma-Aldrich
Moxidectin, ≥98% (HPLC)