Merck
  • Long non‑coding RNA CASC15 facilitates esophageal squamous cell carcinoma tumorigenesis via decreasing SIM2 stability via FTO‑mediated demethylation.
Long non‑coding RNA CASC15 facilitates esophageal squamous cell carcinoma tumorigenesis via decreasing SIM2 stability via FTO‑mediated demethylation.
Oncology reports (2021-03-03)
Bo Qin, Meng Dong, Zhengyang Wang, Jiajia Wan, Yingying Xie, Yi Jiao, Dan Yan
ABSTRACT
Long non‑coding RNAs (lncRNAs) are involved in the regulation of esophageal squamous cell carcinoma (ESCC) progression. However, the function and mechanism of lncRNA cancer susceptibility candidate 15 (CASC15) are poorly defined. In the present study, tumor and normal adjacent tissues were collected from 45 patients with ESCC. Expression levels of CASC15, fat mass and obesity‑associated (FTO) protein and single‑minded 2 (SIM2) were examined via reverse transcription‑quantitative PCR and western blot assays. Cell proliferation and apoptosis were evaluated via MTT, flow cytometry and caspase‑3 activity assays, respectively. Additionally, an ESCC mouse xenograft model was used to assess the function of CASC15 in vivo. The interaction between FTO and CASC15/SIM2 was analyzed via RNA immunoprecipitation and RNA pull‑down assays. The results revealed that CASC15 expression was elevated in ESCC tissues, and patients with ESCC exhibiting high CASC15 expression had a poor prognosis. CASC15‑knockdown inhibited ESCC cell proliferation and facilitated apoptosis. Additionally, CASC15‑knockdown decreased the growth of ESCC xenograft tumors. CASC15 decreased SIM2 stability via FTO‑mediated demethylation. Additionally, FTO loss markedly weakened CASC15‑mediated pro‑proliferative and anti‑apoptotic effects in ESCC cells. SIM2 downregulation weakened the effect of CASC15‑knockdown on cell proliferation and inhibited the increase of the apoptotic rate and caspase‑3 activity induced by CASC15 depletion in ESCC cells. In conclusion, CASC15 promoted ESCC tumorigenesis by decreasing SIM2 stability via FTO‑mediated demethylation.
Sigma-Aldrich
Normal Mouse IgG, Normal Mouse IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
Sigma-Aldrich
Normal Rabbit IgG, Normal Rabbit IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
Sigma-Aldrich
Anti-N6-methyladenosine Antibody (m6A), clone 17-3-4-1, clone 17-3-4-1, from mouse