Merck
  • Decreased Activity of the Ghrhr and Gh Promoters Causes Dominantly Inherited GH Deficiency in Humanized GH1 Mouse Models.
Decreased Activity of the Ghrhr and Gh Promoters Causes Dominantly Inherited GH Deficiency in Humanized GH1 Mouse Models.
Endocrinology (2019-08-23)
Daisuke Ariyasu, Emika Kubo, Daisuke Higa, Shinsuke Shibata, Yutaka Takaoka, Michihiko Sugimoto, Kazunori Imaizumi, Tomonobu Hasegawa, Kimi Araki
ABSTRACT
Isolated growth hormone deficiency type II (IGHD2) is mainly caused by heterozygous splice-site mutations in intron 3 of the GH1 gene. A dominant-negative effect of the mutant GH lacking exon 3 on wild-type GH secretion has been proposed; however, the molecular mechanisms involved are elusive. To uncover the molecular systems underlying GH deficiency in IGHD2, we established IGHD2 model mice, which carry both wild-type and mutant copies of the human GH1 gene, replacing each of the endogenous mouse Gh loci. Our IGHD2 model mice exhibited growth retardation along with intact cellular architecture and mildly activated endoplasmic reticulum stress in the pituitary gland, caused by decreased GH-releasing hormone receptor (Ghrhr) and Gh gene promoter activities. Decreased Ghrhr and Gh promoter activities were likely caused by reduced levels of nuclear CREB3L2, which was demonstrated to stimulate Ghrhr and Gh promoter activity. To our knowledge, this is the first in vivo study to reveal a novel molecular mechanism of GH deficiency in IGHD2, representing a new paradigm that differs from widely accepted models.
Sigma-Aldrich
ApopTag Peroxidase In Situ Apoptosis Detection Kit, The ApopTag Peroxidase In Situ Apoptosis Detection Kit detects apoptotic cells in situ by labeling & detecting DNA strand breaks by the TUNEL method.
Sigma-Aldrich
Anti-OASIS/CREB3L1 Antibody, clone 44C7, clone 44C7, from mouse
Roche
DIG RNA Labeling Kit (SP6/T7), sufficient for 2 x 10 labeling reactions, kit of 1 (12 components), suitable for hybridization, suitable for Southern blotting