Merck
  • Protein-caged zinc porphyrin as a carbonic anhydrase mimic for carbon dioxide capture.
Protein-caged zinc porphyrin as a carbonic anhydrase mimic for carbon dioxide capture.
Scientific reports (2020-11-13)
Haixia Chi, Han Chen, Kai Gong, Xiaoqiang Wang, Youming Zhang
ABSTRACT
Zinc tetraphenylporphyrin (Zn-TPP) solubilized by GroEL protein cage was prepared as a supramolecular mimic of carbonic anhydrase (CA) for CO2 capture. It is shown that the soluble Zn-TPP-GroEL complex can be formed easily by detergent dialysis. The Zn-TPP/GroEL binding ratio was found to increase with their dialysis ratio until reaching the maximum of about 30 porphyrins per protein cage. Moreover, the complex showed hydrase activity that catalyzes the CO2 hydration in HCO3- and H+. It is further seen that the catalytic activity of Zn-TPP-GroEL was about one-half of that of a bovine CA at 25 °C. On the other hand, as the temperature was increased to 60 °C close to an industrial CO2 absorption temperature, the natural enzyme lost function while Zn-TPP-GroEL exhibited better catalytic performance indicative of a higher thermal stability. Finally, we demonstrate that the GroEL-solubilized Zn-TPP is able to accelerate the precipitation of CO2 in the form of CaCO3 and has better long-term performance than the bovine CA. Thus a new type of nano-caged system mimicking natural CAs for potential applications in carbon capture has been established.
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, ≥95% (SDS-PAGE), specific activity ≥3,500 W-A units/mg protein, lyophilized powder